Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009872.3 | NP_034002.2 | 1 | 748 | ||
GenBank mRNA | BC100715.1 | - | 2 | 427 | ||
BC103550.1 | - | 2 | 410 |