Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010371.2 | NP_034501.2 | 3 | 382 | ||
GenBank mRNA | BC120755.1 | - | 3 | 332 | 40129678 | |
BC120757.1 | - | 3 | 332 | 40129680 | ||
BC145030.1 | - | 3 | 332 | 9053578 | ||
M18459.1 | - | 3 | 382 | |||
X12822.1 | - | 3 | 326 |