Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008213.2 | NP_032239.1 | 1 | 379 | ||
GenBank mRNA | AK146084.1 | - | 280 | |||
BC050182.1 | - | 1 | 229 | 30022934 | ||
S79216.1 | - | 1 | 338 | |||
U21226.1 | - | 1 | 379 |