Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010498.3 | NP_034628.2 | 9 | 1540 | ||
GenBank mRNA | AJ627044.1 | - | 7 | 1005 | ||
AK085237.1 | - | 10 | 1417 | |||
AK132287.1 | - | 10 | 1420 | |||
AK166178.1 | - | 9 | 1418 | |||
AK166443.1 | - | 10 | 1369 | |||
L07921.1 | - | 9 | 1416 |