Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhbe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008382.2 | NP_032408.2 | 2 | 1149 | ||
GenBank mRNA | BC010404.1 | - | 2 | 990 | 4160980 | |
U96386.1 | - | 2 | 1137 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094452.1 | - | 2423 | Chr. - - [-] on Build |