Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021557.5 | NP_067532.2 | 5 | 571 | ||
GenBank mRNA | AB035959.1 | - | 5 | 579 | ||
AF474027.1 | - | 5 | 617 | |||
AK004413.1 | - | 5 | 572 | |||
AK012056.1 | - | 5 | 599 | |||
AK135443.1 | - | 5 | 601 | |||
AY039032.1 | - | 5 | 1045 | |||
BC018261.1 | - | 5 | 596 | 4460226 | ||
BC036273.1 | - | 5 | 596 |