Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029855.1 | NP_001025026.1 | 8 | 484 | ||
NM_011190.3 | NP_035320.1 | 8 | 584 | |||
GenBank mRNA | AA267668.1 | - | 8 | 475 | ||
AK012344.1 | - | 8 | 513 | |||
AK088654.1 | - | 8 | 511 | |||
AK143556.1 | - | 8 | 516 | |||
BC005680.1 | - | 8 | 504 | 3154255 | ||
BC057859.1 | - | 8 | 581 | 5310526 | ||
BY498670.1 | - | 2 | 90 | |||
CA536627.1 | - | 8 | 496 |
Gene Symbol: Psme2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281472.1 | NP_001268401.1 | 1 | 744 | ||
GenBank mRNA | D87910.1 | - | 1 | 471 |