Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109992.1 | NP_001103462.1 | 3, 4 | 515 | ||
NM_011202.3 | NP_035332.1 | 3, 4 | 515 | |||
GenBank mRNA | AK011468.1 | - | 3, 4 | 477 | ||
AK147354.1 | - | 3, 4 | 486 | |||
AK159501.1 | - | 3, 4 | 515 | |||
AK159587.1 | - | 3, 4 | 515 | |||
BC028513.1 | - | 3, 4 | 465 | |||
BC057398.1 | - | 3, 4 | 447 | 6409640 | ||
BC059278.1 | - | 3, 4 | 493 | 2866038 | ||
D84372.1 | - | 3, 4 | 391 |