Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159525.1 | NP_001152997.1 | 4 | 478 | ||
NM_023041.3 | NP_075528.3 | 6 | 754 | |||
GenBank mRNA | AK012785.1 | - | 6 | 737 | ||
AK028449.1 | - | 4 | 472 | |||
AK029368.1 | - | 6 | 755 | |||
AK075576.1 | - | 6 | 741 | |||
AK170175.1 | - | 6 | 743 | |||
AK210467.1 | - | 1 | 114 | |||
AK216660.1 | - | 1 | 132 | |||
AK218946.1 | - | 3 | 336 | |||
BC012517.1 | - | 6 | 689 | |||
BC019767.1 | - | 6 | 737 | 4502819 | ||
CT010305.1 | - | 6 | 730 | |||
Y09046.1 | - | 6 | 735 |