Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293652.1 | NP_001280581.1 | 8 | 1036 | ||
NM_001293653.1 | NP_001280582.1 | 6 | 697 | |||
NM_001293654.1 | NP_001280583.1 | 7 | 863 | |||
NM_001293655.1 | NP_001280584.1 | 6 | 776 | |||
NM_009005.3 | NP_033031.2 | 6 | 814 | |||
GenBank mRNA | AB232599.1 | - | 5 | 608 | ||
AK005004.1 | - | 6 | 776 | |||
AK057671.1 | - | 6 | 743 | |||
AK135659.1 | - | 7 | 865 | |||
AK139914.1 | - | 6 | 767 | |||
AK148117.1 | - | 6 | 807 | |||
AK151078.1 | - | 6 | 788 | |||
AK152666.1 | - | 8 | 1011 | |||
AK154234.1 | - | 6 | 795 | |||
AK158971.1 | - | 6 | 698 | |||
AK159507.1 | - | 6 | 743 | |||
AK159928.1 | - | 6 | 715 | |||
BC004597.1 | - | 6 | 732 | 3584891 | ||
BC086793.1 | - | 6 | 730 | 6414012 | ||
CT010352.1 | - | 5 | 608 | |||
X89650.1 | - | 6 | 764 |