Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177792.1 | NP_001171263.1 | 7, 8 | 621 | ||
NM_001177793.1 | NP_001171264.1 | 6, 7 | 543 | |||
NM_009222.3 | NP_033248.1 | 6, 7 | 588 | |||
GenBank mRNA | AB000822.1 | - | 5, 6 | 463 | ||
AB007444.1 | - | 6, 7 | 520 | |||
AF007169.1 | - | 5, 6 | 412 | |||
AK017311.1 | - | 7, 8 | 596 | |||
AK019162.1 | - | 6, 7 | 589 | |||
BC070456.1 | - | 6, 7 | 493 | 30606358 | ||
U73143.1 | - | 5, 6 | 412 |