Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144820.3 | NP_659069.2 | 4, 5 | 641 | ||
GenBank mRNA | AK148447.1 | - | 4, 5 | 642 | ||
BC027759.1 | - | 4, 5 | 649 | 5346317 |