Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilvbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173751.4 | NP_776112.1 | 11 | 1433 | ||
GenBank mRNA | AK030933.1 | - | 8 | 871 | ||
AK087956.1 | - | 10 | 1307 | |||
AK146075.1 | - | 10 | 1361 | |||
AK169899.1 | - | 10 | 1359 | |||
AK184668.1 | - | 2 | 139 | |||
BC052054.1 | - | 10 | 1767 | |||
BC056459.1 | - | 11 | 1391 | 5718157 |