Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009394.2 | NP_033420.1 | 4, 5 | 389 | ||
GenBank mRNA | AK133548.1 | - | 4, 5 | 385 | ||
BC024390.1 | - | 4, 5 | 388 | 5026329 |