Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159392.1 | NP_001152864.1 | 5, 6 | 818 | ||
NM_009395.4 | NP_033421.3 | 5, 6 | 831 | |||
GenBank mRNA | AK004593.2 | - | 5, 6 | 745 | ||
AK034305.1 | - | 5, 6 | 766 | |||
AK147809.1 | - | 5, 6 | 726 | |||
AK168386.1 | - | 5, 6 | 710 | |||
AK168704.1 | - | 5, 6 | 753 | |||
AK168805.1 | - | 5, 6 | 726 | |||
BC003906.1 | - | 5, 6 | 667 | 3484017 |