Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013697.5 | NP_038725.1 | 1 | 222 | ||
GenBank mRNA | AK014454.1 | - | 1 | 59 | ||
AK018701.1 | - | 1 | 60 | |||
AK050155.1 | - | 1 | 59 | |||
AK075588.1 | - | 1 | 59 | |||
BC024702.1 | - | 1 | 192 | 4192268 | ||
BC032069.1 | - | 1 | 49 | |||
BC086926.1 | - | 1 | 59 | 6749152 | ||
CK627038.1 | - | 1 | 48 | |||
D89076.1 | - | 1 | 59 | |||
X03351.1 | - | 1 | 59 |