Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C030006K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145472.2 | NP_663447.1 | 1 | 56 | ||
NM_176828.4 | NP_789798.2 | 1 | 56 | |||
GenBank mRNA | AK021050.1 | - | 1 | 45 | ||
AK046662.1 | - | 1 | 24 | |||
AK053546.1 | - | 1 | 34 | |||
AK075645.1 | - | 1 | 52 | |||
AK085641.1 | - | 1 | 52 | |||
AK140368.1 | - | 1 | 52 | |||
AK140578.1 | - | 1 | 57 | |||
AK163615.1 | - | 1 | 39 | |||
AK165937.1 | - | 1 | 50 | |||
BC019462.1 | - | 1 | 32 | 4458439 | ||
BC026997.1 | - | 1 | 30 | 5362226 |