Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301705.1 | NP_001288634.1 | 4 | 651 | ||
NM_144899.3 | NP_659148.2 | 4 | 533 | |||
GenBank mRNA | AK155481.1 | - | 4 | 581 | ||
AY158701.1 | - | 4 | 474 | |||
BC117925.1 | - | 3 | 446 | 40097608 |