Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hykk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177351.4 | NP_796325.2 | 5 | 1578 | ||
GenBank mRNA | AK048771.1 | - | 4 | 1411 | ||
AK085527.1 | - | 5 | 1587 | |||
AK138851.1 | - | 5 | 1564 | |||
AK162425.1 | - | 5 | 1579 | |||
BC038618.1 | - | 5 | 1557 | 4221933 |