Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013381.2 | NP_001013399.1 | 3 | 536 | ||
GenBank mRNA | AK046706.1 | - | 3 | 541 | ||
AK148401.1 | - | 3 | 531 | |||
BB645699.1 | - | 3 | 541 | |||
BC139274.1 | - | 3 | 479 | 8862296 | ||
BC139276.1 | - | 3 | 479 | 8862298 |