Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1ql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024702.1 | NP_001019873.1 | 2 | 561 | ||
GenBank mRNA | BC147036.1 | - | 2 | 650 | 9056046 | |
BC147037.1 | - | 2 | 650 | 9056067 | ||
DQ002403.1 | - | 2 | 561 |