Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026244.2 | NP_080520.2 | 3 | 931 | ||
GenBank mRNA | AK014732.1 | - | 3 | 637 | ||
AK014734.1 | - | 3 | 637 | |||
AK080462.1 | - | 3 | 913 | |||
AK090062.1 | - | 3 | 924 | |||
BC043112.1 | - | 3 | 878 | |||
BC058649.1 | - | 3 | 1263 | |||
BC072655.1 | - | 3 | 907 | |||
BC117754.1 | - | 3 | 525 | 40090588 | ||
BC117755.1 | - | 3 | 525 | 40090589 |