Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175171.3 | NP_780380.2 | 9 | 1389 | ||
GenBank mRNA | AK090136.1 | - | 9 | 1361 |