Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183208.3 | NP_899031.2 | 7, 8 | 1021 | ||
GenBank mRNA | AK054366.1 | - | 7, 8 | 915 | ||
BC058646.1 | - | 7, 8 | 939 | 6405777 | ||
BC065120.1 | - | 7, 8 | 914 | 6856060 |