Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163358.1 | NP_001156830.1 | 3, 4 | 165 | ||
NM_178247.3 | NP_839978.1 | 4, 5 | 251 | |||
GenBank mRNA | AF490345.1 | - | 4, 5 | 237 | ||
AK145642.1 | - | 4, 5 | 252 | |||
AK166752.1 | - | 4, 5 | 249 | |||
BC115806.1 | - | 3, 4 | 192 | 40106955 | ||
BC115901.1 | - | 3, 4 | 189 | 40106956 |