Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198808.1 | NP_001185737.1 | 3 | 276 | ||
NM_001198809.1 | NP_001185738.1 | 3 | 355 | |||
NM_177386.5 | NP_796360.2 | 4 | 399 | |||
GenBank mRNA | AK040600.1 | - | 3 | 356 | ||
AK049505.1 | - | 3 | 218 | |||
AK052339.1 | - | 3 | 342 | |||
AK147557.1 | - | 3 | 281 | |||
AK147651.1 | - | 4 | 254 | |||
AK168444.1 | - | 5 | 304 | |||
AK220408.1 | - | 1 | 141 | |||
BB636293.1 | - | 3 | 356 | |||
BC115510.1 | - | 3 | 210 | |||
BC115511.1 | - | 3 | 210 | |||
BC139354.1 | - | 3 | 187 | 8862376 | ||
BC145610.1 | - | 2 | 143 | 9054176 |