Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290837.1 | NP_001277766.1 | 2 | 289 | ||
NM_011354.3 | NP_035484.1 | 2 | 289 | |||
GenBank mRNA | AF073297.1 | - | 2 | 70 | ||
AK002351.1 | - | 2 | 65 | |||
AK008802.1 | - | 2 | 78 | |||
AK010749.1 | - | 2 | 121 | |||
AK012395.1 | - | 2 | 82 | |||
AK012603.1 | - | 2 | 68 | |||
AK018746.1 | - | 2 | 65 | |||
AK132746.1 | - | 2 | 82 | |||
AK151362.1 | - | 2 | 82 | |||
AK168858.1 | - | 2 | 69 | |||
AK169106.1 | - | 2 | 71 | |||
BC081455.1 | - | 2 | 73 | |||
BY070249.1 | - | 2 | 105 | |||
CJ073462.1 | - | 2 | 295 |
Gene Symbol: Fam221b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038644.1 | - | 3 | 2995 |