Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025083.2 | NP_001020254.1 | 4 | 781 | ||
GenBank mRNA | BC132522.1 | - | 4 | 776 | 40130799 | |
BC132548.1 | - | 4 | 776 | 40130825 | ||
BU757254.1 | - | 1 | 167 | |||
DQ058649.1 | - | 4 | 781 |