Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098789.1 | NP_001092259.1 | 1 | 60 | ||
GenBank mRNA | AK131609.1 | - | 1 | 32 | ||
AK131662.1 | - | 1 | 29 | |||
BC064011.1 | - | 1 | 21 | |||
BC099496.1 | - | 1 | 21 | 30847512 | ||
BY336656.1 | - | 1 | 62 |