Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028763.3 | NP_083039.2 | 5 | 804 | ||
GenBank mRNA | AK004679.2 | - | 5 | 709 | ||
AK035250.1 | - | 5 | 711 | |||
AK090253.1 | - | 5 | 657 | |||
AK146048.1 | - | 5 | 655 | |||
AK146103.1 | - | 5 | 655 | |||
AK147702.1 | - | 1 | 5407 | |||
BC011199.1 | - | 5 | 693 | |||
BC048240.1 | - | 5 | 701 | 6493113 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY395907.1 | - | 5 | 709 |