Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026383.3 | NP_080659.1 | 3 | 450 | ||
GenBank mRNA | AK011192.1 | - | 3 | 428 | ||
AK012073.1 | - | 3 | 434 | |||
AK013163.1 | - | 3 | 434 | |||
AK018384.1 | - | 3 | 398 | |||
AK031048.1 | - | 1448 | ||||
AK075816.1 | - | 3 | 434 | |||
AK077403.1 | - | 3 | 315 | |||
AK207101.1 | - | 1 | 46 | |||
AK207137.1 | - | 1 | 46 | |||
AK210203.1 | - | 1 | 46 | |||
AK215675.1 | - | 1 | 46 | |||
AK218125.1 | - | 2 | 289 | |||
BC006598.1 | - | 3 | 423 | 3965195 |