Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 12 | 1562 | ||
GenBank mRNA | AB028921.1 | - | 12 | 1558 | ||
AK011325.1 | - | 12 | 1567 | |||
AK166526.1 | - | 13 | 1478 | |||
BC003242.1 | - | 2 | 191 | |||
BC046801.1 | - | 12 | 1551 | 6403966 | ||
BC051015.1 | - | 10 | 3871 |