Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mansc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034903.3 | NP_001030075.1 | 3 | 634 | ||
GenBank mRNA | AK138652.1 | - | 3 | 635 |