Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033793.3 | NP_001028965.1 | 3 | 1470 | ||
GenBank mRNA | AK139937.1 | - | 3 | 1471 | ||
AK163257.1 | - | 2 | 1587 | |||
BC139141.1 | - | 2 | 1285 | 8862163 | ||
BC145822.1 | - | 2 | 1285 | 40131125 |