Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021718.2 | NP_068364.1 | 2 | 177 | ||
GenBank mRNA | AF230640.1 | - | 2 | 134 | ||
AF237913.1 | - | 2 | 146 | |||
AF354929.1 | - | 2 | 134 | |||
AK039824.1 | - | 2 | 152 | |||
AK080934.1 | - | 2 | 178 | |||
AK088631.1 | - | 2 | 134 | |||
AK088780.1 | - | 2 | 183 | |||
AK138737.1 | - | 2 | 150 | |||
AK172495.1 | - | 2 | 150 | |||
BC028433.1 | - | 2 | 134 | 1248222 |