Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023179.3 | NP_075668.1 | 2 | 294 | ||
GenBank mRNA | AB076405.1 | - | 2 | 349 | ||
AK005109.1 | - | 2 | 175 | |||
AK011945.1 | - | 2 | 137 | |||
BC020190.1 | - | 2 | 290 | 4505711 | ||
BC025190.1 | - | 283 | ||||
BC049587.1 | - | 2 | 159 | |||
BC062380.1 | - | 2 | 282 | 6468413 | ||
BU558667.1 | - | 2 | 297 |