Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 4 | 731 | ||
NM_023210.4 | NP_075699.3 | 4 | 731 | |||
GenBank mRNA | AB037685.1 | - | 4 | 406 | ||
AK012759.1 | - | 4 | 698 | |||
AK049647.1 | - | 4 | 698 | |||
AK076049.1 | - | 4 | 558 | |||
AK088401.1 | - | 4 | 711 | |||
AK147888.1 | - | 4 | 697 | |||
AK159446.1 | - | 4 | 698 | |||
AK168380.1 | - | 4 | 680 | |||
AK172146.1 | - | 4 | 745 | |||
AK193570.1 | - | 2 | 193 | |||
AK195817.1 | - | 2 | 102 | |||
AK199406.1 | - | 2 | 193 | |||
AK202251.1 | - | 2 | 195 | |||
AK204175.1 | - | 2 | 193 | |||
BC005690.1 | - | 4 | 666 | 3596597 | ||
BC080684.1 | - | 4 | 671 | 6493476 | ||
CF952181.1 | - | 4 | 402 | |||
U89345.2 | - | 4 | 692 |