Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025902.3 | NP_080178.1 | 3 | 404 | ||
GenBank mRNA | AK003486.1 | - | 3 | 375 | ||
AK004257.1 | - | 3 | 375 | |||
AK005184.1 | - | 3 | 363 | |||
AM162549.1 | - | 3 | 896 | |||
BC058279.1 | - | 3 | 404 | 6528467 |