Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133687.2 | NP_598448.1 | 2 | 503 | ||
GenBank mRNA | AK005364.1 | - | 2 | 532 | ||
AK015150.1 | - | 2 | 407 | |||
AK132950.1 | - | 2 | 427 | |||
AK191595.1 | - | 1 | 60 | |||
BC016207.1 | - | 1 | 194 | 4912395 | ||
BC089314.1 | - | 2 | 503 | 30541251 |