Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026485.2 | NP_080761.2 | 4 | 339 | ||
GenBank mRNA | AK005300.1 | - | 4 | 309 | ||
AK017745.1 | - | 4 | 333 | |||
AK146027.1 | - | 4 | 355 | |||
AK186431.1 | - | 1 | 46 | |||
BC005574.1 | - | 4 | 365 | 3967961 | ||
BU510197.1 | - | 4 | 419 | |||
CX200611.1 | - | 4 | 324 |