Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176832.2 | NP_789802.1 | 14 | 1569 | ||
NM_194355.2 | NP_919336.1 | 15 | 1841 | |||
GenBank mRNA | BC046486.1 | - | 8 | 924 | ||
BC058669.1 | - | 15 | 1709 | |||
BC079633.1 | - | 5 | 417 | |||
BC094375.1 | - | 15 | 1762 | 30535423 |