Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009573.2 | NP_001009573.2 | 11 | 947 | ||
GenBank mRNA | AK220199.1 | - | 4 | 255 | ||
BC141289.1 | - | 10 | 859 | 9055901 | ||
CF164660.1 | - | 6 | 471 | |||
EF127645.1 | - | 10 | 895 |