Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162979.1 | NP_001156451.1 | 9 | 1425 | ||
GenBank mRNA | AK014876.1 | - | 9 | 1408 | ||
AK029544.1 | - | 9 | 1424 | |||
BC127049.1 | - | 9 | 1412 | |||
BC152307.1 | - | 9 | 1414 | 40062409 | ||
BC152308.1 | - | 9 | 1414 | 40062416 |