Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023644.4 | NP_076133.3 | 6 | 645 | ||
GenBank mRNA | AF310338.1 | - | 6 | 566 | ||
AK007782.1 | - | 6 | 598 | |||
AK031072.1 | - | 6 | 606 | |||
AK032203.1 | - | 6 | 577 | |||
AK160184.1 | - | 4 | 362 | |||
AK168589.1 | - | 6 | 534 | |||
BC021382.1 | - | 6 | 609 | 5042993 |