Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028351.3 | NP_082627.3 | 2 | 804 | ||
GenBank mRNA | AB055811.1 | - | 2 | 762 | ||
AK013366.1 | - | 2 | 753 | |||
AK076308.1 | - | 751 | ||||
AK133916.1 | - | 2 | 805 | |||
AY864332.1 | - | 2 | 265 | |||
BC103794.1 | - | 2 | 738 | 6517974 | ||
BC145929.1 | - | 2 | 409 | 40131232 | ||
BC145931.1 | - | 2 | 409 | 40131234 |