Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175235.3 | NP_780444.2 | 8 | 1033 | ||
GenBank mRNA | AK018246.1 | - | 7 | 826 | ||
AK034216.1 | - | 7 | 829 | |||
BC052406.1 | - | 8 | 1033 | 5716119 | ||
BC057083.1 | - | 7 | 815 | 5707546 |