Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029821.2 | NP_084097.1 | 1, 2 | 247 | ||
GenBank mRNA | AK004470.1 | - | 1, 2 | 465 | ||
AK013117.1 | - | 1, 2 | 249 | |||
BC051545.1 | - | 1, 2 | 415 | 5255010 | ||
DQ225768.1 | - | 1, 2 | 122 |