Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330007P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029951.1 | NP_084227.1 | 7 | 768 | ||
GenBank mRNA | AK032304.1 | - | 7 | 683 | ||
AK034943.1 | - | 7 | 709 | |||
AK035825.1 | - | 7 | 757 | |||
AK036910.1 | - | 7 | 691 | |||
AK044209.1 | - | 7 | 705 | |||
AK141615.1 | - | 7 | 709 | |||
BC021479.1 | - | 913 | 5344198 | |||
BC051144.1 | - | 7 | 681 | 6588980 | ||
BC085302.1 | - | 7 | 656 |