Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030699.2 | NP_109624.1 | 6 | 2308 | ||
GenBank mRNA | AB038667.1 | - | 6 | 2085 | ||
AF475069.1 | - | 5 | 1213 | |||
AF475075.1 | - | 5 | 1213 | |||
AF475076.1 | - | 5 | 1213 | |||
AK039260.1 | - | 1 | 750 | |||
AK048796.1 | - | 5 | 1174 | |||
AK134467.1 | - | 5 | 2138 |