Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271725.1 | NP_001258654.1 | 6 | 1436 | ||
NM_001271726.1 | NP_001258655.1 | 6 | 1472 | |||
NM_001271727.1 | NP_001258656.1 | 6 | 1741 | |||
NM_001271728.1 | NP_001258657.1 | 6 | 1539 | |||
NM_030706.3 | NP_109631.2 | 6 | 1535 | |||
GenBank mRNA | AB043550.1 | - | 6 | 1519 | ||
AF220017.1 | - | 6 | 1384 | |||
AK039741.1 | - | 6 | 1533 | |||
AK045410.1 | - | 6 | 1532 | |||
AK079415.1 | - | 6 | 1686 | |||
AK141419.1 | - | 6 | 1382 | |||
AK147275.1 | - | 6 | 1649 | |||
AK147395.1 | - | 6 | 1530 | |||
AK164350.1 | - | 5 | 1238 | |||
AK220343.1 | - | 6 | 1740 | |||
BC058668.1 | - | 5 | 2347 | |||
BC058961.1 | - | 6 | 1539 | 6826143 |